Restek™ Replacement Frits for High-Pressure Frit-Type In-Line Filters
Replacement guard cartridges can cost as much as an analytical column, so why not protect them, too? The removable cap frit filter in a Trident direct helps prevent clogged cartridges to extend the life of your column, your cartridge, and your budget.
Supplier: Restek™ RK25022
Description
- Replacement frits for use with Restek High-Pressure Frit-Type In-Line Filters
- Use 2mm cap frit filters with 1.0-2.1 mm ID analytical columns
- Use 4mm cap frit filters with 3.0-4.6mm ID analytical columns
Specifications
Replacement Cap Frits | |
4 mm | |
2 μm | |
5 Pack |