Restek™ Replacement Frits for High-Pressure Frit-Type In-Line Filters
Replacement guard cartridges can cost as much as an analytical column, so why not protect them, too? The removable cap frit filter in a Trident direct helps prevent clogged cartridges to extend the life of your column, your cartridge, and your budget.
$161.87 - $163.51
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Porosity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Porosity | Price | Quantity | ||||
06712631
|
Restek™
RK25057 |
2 mm | 2 μm |
Each for $161.87
|
|
||||
06719062
|
Restek™
RK25022 |
4 mm | 2 μm |
Pack of 5 Each for $163.51
|
|
||||
06719063
|
Restek™
RK25023 |
4 mm | 0.5 μm |
Pack of 5 Each for $161.87
|
|
||||
Description
- Replacement frits for use with Restek High-Pressure Frit-Type In-Line Filters
- Use 2mm cap frit filters with 1.0-2.1 mm ID analytical columns
- Use 4mm cap frit filters with 3.0-4.6mm ID analytical columns
Specifications
Replacement Cap Frits |