Restek™ Replacement Frits for High-Pressure Frit-Type In-Line Filters DFS Item
Replacement guard cartridges can cost as much as an analytical column, so why not protect them, too? The removable cap frit filter in a Trident direct helps prevent clogged cartridges to extend the life of your column, your cartridge, and your budget.
Supplier:  Restek™ RK25023
 View more versions of this product
Catalog No. 06719063
Description
Description
  • Replacement frits for use with Restek High-Pressure Frit-Type In-Line Filters
  • Use 2mm cap frit filters with 1.0-2.1 mm ID analytical columns
  • Use 4mm cap frit filters with 3.0-4.6mm ID analytical columns
Specifications
Specifications
Replacement Cap Frits
4 mm
0.5 μm
5 Pack