Restek™ Rxi™-XLB GC Columns DFS Item
General-purpose columns exhibiting extremely low bleed
Supplier:  Restek™ RK43702
 View more versions of this product
Catalog No. 06-802-965
Explore available promotions
Description
Description
  • Especially well-suited for analysis of active, high molecular weight compounds with sensitive GC/MS systems including ion trap detectors
  • Temperature range 30° to 360°C for 15m and 30m lengths: longer columns may have a slightly reduced maximum temperature
  • Well-suited for U.S. EPA Method 525 for analysis of semivolatile compounds in drinking water
Pesticides, PCB cogeners
Specifications
Specifications
0.18 mm
Pesticide Analysis, PCB Congener Analysis, PAHs Analysis
30°C to 340/360°C
20 m
Fused Silica
Rxi™-XLB
0.18 μm
0.18 μm
Low polar