Restek™ Rxi™-XLB GC Columns
General-purpose columns exhibiting extremely low bleed
$1409.39 - $4016.50
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity | |||
06-802-927
|
Restek™
RK13723 |
0.25 mm | 0.25 μm | 30 m |
Each for $1,663.40
|
|
|||
06-802-955
|
Restek™
RK13754 |
0.32 mm | 1 μm | 30 m |
Each for $1,823.49
|
|
|||
06-802-948
|
Restek™
RK13705 |
0.25 mm | 0.1 μm | 15 m |
Each for $1,409.39
|
|
|||
06-802-949
|
Restek™
RK13708 |
0.25 mm | 0.1 μm | 30 m |
Each for $2,320.00
|
|
|||
06-802-951
|
Restek™
RK13726 |
0.25 mm | 0.25 μm | 60 m |
Each for $4,016.50
|
|
|||
06-802-930
|
Restek™
RK13738 |
0.25 mm | 0.5 μm | 30 m |
Each for $1,748.64
|
|
|||
06-802-952
|
Restek™
RK13740 |
0.53 mm | 0.5 μm | 30 m |
Each for $2,025.19
|
|
|||
06-802-965
|
Restek™
RK43702 |
0.18 mm | 0.18 μm | 20 m |
Each for $1,507.45
|
|
|||
Description
- Especially well-suited for analysis of active, high molecular weight compounds with sensitive GC/MS systems including ion trap detectors
- Temperature range 30° to 360°C for 15m and 30m lengths: longer columns may have a slightly reduced maximum temperature
- Well-suited for U.S. EPA Method 525 for analysis of semivolatile compounds in drinking water
Pesticides, PCB cogeners
Specifications
Fused Silica |