Restek™ Valco Connectors DFS Item
For use with 0.0625 in. OD tubing.
Supplier:  Restek™ RK20148
 View more versions of this product
Catalog No. 06710515
Explore available promotions
Description
Description
  • Ends of tubing seat squarely at bottoms of fitting details
  • Made of 300-series stainless steel
Specifications
Specifications
Union
0.25 mm
HPLC