missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Restore™ Western Blot Stripping Buffer

Safely and effectively remove primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.

$87.58 - $1330.77

Specifications

Form Liquid
Product Line Restore™
Content And Storage Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature.
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FER21059X4
View Documents
Thermo Scientific™
21059X4
4 x 500 mL
Each for $1,142.59
 
PI21062
View Documents
Thermo Scientific™
LSG21062
30 mL
Each for $87.58
 
PI21059
View Documents
Thermo Scientific™
LSG21059
500 mL
Each for $399.91
 
PI21063
View Documents
Thermo Scientific™
LSG21063
5 L
Each for $1,330.77
 
Description

Description

Thermo Scientific™ Restore Western Blot Stripping Buffer safely and effectively removes primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.

Performing gel electrophoresis and duplicate immunoblot assays to test new primary antibodies or antibody concentrations is time-consuming and expensive. Restore Western Blot Stripping Buffer eliminates this waste when detecting immunoblots with chemiluminescent Western blotting substrates. The reagent allows clean and efficient removal of primary and secondary antibodies from immunoblots without removing or damaging the immobilized antigen. This allows blots to be stripped and reprobed with confidence.

Highlights:

  • Saves time – no need to re-run gels and blots

  • Saves costly sample – re-probe the membrane using the same target sample

  • Effective – formulation is more efficient at stripping antibodies than homemade buffers

  • Gentle – does not damage the target antigen during stripping allowing efficient reprobing

  • Odor-free – no mercaptans means no acrid odors

  • Economical – less expensive than other commercial stripping buffers

Reuse a nitrocellulose or PVDF blot to detect a different target with a different primary antibody; Reprobe a blot to correct or optimize antibody concentrations that were ineffective the first time.

Specifications

Specifications

Liquid
Restore™
Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.