Thermo Scientific™ Restore™ Western Blot Stripping Buffer
Safely and effectively remove primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
Supplier:  Thermo Scientific™ LSG21062
 View more versions of this product
Catalog No. PI21062
Explore available promotions
Description
Description
Thermo Scientific™ Restore Western Blot Stripping Buffer safely and effectively removes primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
Performing gel electrophoresis and duplicate immunoblot assays to test new primary antibodies or antibody concentrations is time-consuming and expensive. Restore Western Blot Stripping Buffer eliminates this waste when detecting immunoblots with chemiluminescent Western blotting substrates. The reagent allows clean and efficient removal of primary and secondary antibodies from immunoblots without removing or damaging the immobilized antigen. This allows blots to be stripped and reprobed with confidence.
Highlights:
  • Saves time – no need to re-run gels and blots
  • Saves costly sample – re-probe the membrane using the same target sample
  • Effective – formulation is more efficient at stripping antibodies than homemade buffers
  • Gentle – does not damage the target antigen during stripping allowing efficient reprobing
  • Odor-free – no mercaptans means no acrid odors
  • Economical – less expensive than other commercial stripping buffers
Reuse a nitrocellulose or PVDF blot to detect a different target with a different primary antibody; Reprobe a blot to correct or optimize antibody concentrations that were ineffective the first time.
Specifications
Specifications
Western Blot Stripping Buffer
Liquid
Nitrocellulose, PVDF
30 mL
Chemiluminescence
Western Blot
Restore™
Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature.
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.