missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Rhodamine 110, 98+%
Supplier: Thermo Scientific Chemicals 419075000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Rhodamine 110 | |
13558-31-1 | |
C20H15ClN2O3 | |
Solubility in water: slightly soluble. | |
Authentic | |
2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride | |
11617572 | |
366.79 | |
Pure | |
Glass bottle | |
White to Yellow | |
Crystalline Powder |
>360.0°C | |
98+% | |
520nm (in methanol) | |
JNGRENQDBKMCCR-UHFFFAOYSA-N | |
C1=CC=C(C(=C1)C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N)C(=O)O.Cl | |
366.801 | |
498 to 504nm (c=10mg/l in methanol) | |
≥98% | |
77000 l/(mol.cm) (at lambda max.) min. | |
(for use as a laser dye) passes | |
500 mg |
Chemical Identifiers
13558-31-1 | |
366.801 | |
11617572 | |
C1=CC=C(C(=C1)C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N)C(=O)O.Cl |
C20H15ClN2O3 | |
JNGRENQDBKMCCR-UHFFFAOYSA-N | |
2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride |
RUO – Research Use Only