missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Rhodamine 110, 98+%, pure, laser grade
CAS: 13558-31-1 | C20H15ClN2O3 | 366.801 g/mol
$325.40 - $325.40
Chemical Identifiers
| CAS | 13558-31-1 |
|---|---|
| Molecular Formula | C20H15ClN2O3 |
| Molecular Weight (g/mol) | 366.801 |
| InChI Key | JNGRENQDBKMCCR-UHFFFAOYSA-N |
| PubChem CID | 11617572 |
| IUPAC Name | 2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride |
| SMILES | C1=CC=C(C(=C1)C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N)C(=O)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC419075000
|
Thermo Scientific Chemicals
419075000 |
500 mg | Glass bottle |
Each for $325.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13558-31-1 | |
| 366.801 | |
| 11617572 | |
| C1=CC=C(C(=C1)C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N)C(=O)O.Cl |
| C20H15ClN2O3 | |
| JNGRENQDBKMCCR-UHFFFAOYSA-N | |
| 2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride |
Specifications
| >360.0°C | |
| 98+% | |
| 520nm (in methanol) | |
| JNGRENQDBKMCCR-UHFFFAOYSA-N | |
| C1=CC=C(C(=C1)C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N)C(=O)O.Cl | |
| 366.801 | |
| 498 to 504nm (c=10mg/l in methanol) | |
| ≥98% | |
| 77000 l/(mol.cm) (at lambda max.) min. | |
| (for use as a laser dye) passes | |
| 500 mg | |
| Rhodamine 110 |
| 13558-31-1 | |
| C20H15ClN2O3 | |
| Solubility in water: slightly soluble. | |
| Authentic | |
| 2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride | |
| 11617572 | |
| 366.79 | |
| Pure | |
| Glass bottle | |
| White to Yellow | |
| Crystalline Powder |
RUO – Research Use Only