Learn More
Rhodamine B base, 97%, Thermo Scientific Chemicals
Rhodamine B base, CAS # 509-34-2, is a fluorescent dye used in staining applications.
Supplier: Thermo Scientific Chemicals 215231000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Rhodamine B base | |
99% | |
MFCD00066967 | |
C.I. 45170.1, Solvent Red 49 | |
Authentic | |
442.55 | |
Glass bottle | |
100 g |
509-34-2 | |
C28H30N2O3 | |
19, 344 | |
Solubility in water: insoluble. Other solubilities: soluble in chloroform, dimethylformamide,, dimethylsulfoxide, ethanol | |
1% max. (105°C) | |
97% | |
Lilac | |
Fine Crystalline Powder |
Chemical Identifiers
509-34-2 | |
C28H30N2O3 | |
MFCD00066967 | |
C.I. 45170.1, Solvent Red 49 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Suspected of causing genetic defects.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only