missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodamine B base, 97%, Thermo Scientific Chemicals

Rhodamine B base, CAS # 509-34-2, is a fluorescent dye used in staining applications.

$85.21 - $235.42

Chemical Identifiers

CAS 509-34-2
Molecular Formula C28H30N2O3
MDL Number MFCD00066967
Synonym C.I. 45170.1, Solvent Red 49
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC215230250
SDS View Documents
Thermo Scientific Chemicals
215230250
25 g Glass bottle
Each for $85.21
 
AC215231000
SDS View Documents
Thermo Scientific Chemicals
215231000
100 g Glass bottle
Each for $235.42
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
Rhodamine B is an organic water-soluble fluorescent dye
Rhodamine B has been found as a toxic pollutant in aqueous systems that may be removed by photocatalysis methods
Applications
Rhodamine B has been used to prepare fluorescent probes for iron detection
It can be used for fluorescent labeling of nanoparticles and subsequent detection of nanoparticle uptake in cells
Is used as a dye in ELISA, fluorescence microscopy, and flow cytometry
Specifications

Chemical Identifiers

509-34-2
C28H30N2O3
MFCD00066967
C.I. 45170.1, Solvent Red 49

Specifications

509-34-2
C28H30N2O3
19, 344
Solubility in water: insoluble. Other solubilities: soluble in chloroform, dimethylformamide,, dimethylsulfoxide, ethanol
1% max. (105°C)
97%
Lilac
Fine Crystalline Powder
99%
MFCD00066967
C.I. 45170.1, Solvent Red 49
Authentic
442.55
Glass bottle
25 g
Rhodamine B base
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Suspected of causing genetic defects.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only