missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Riboprobe™ Systems
The Riboprobe Systems are designed to prepare high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts.
$324.13 - $324.13
Specifications
Content And Storage | Store at -20°C. |
---|---|
For Use With (Application) | Generation of high-specific-activity RNA probes, generation of microgram quantities of RNA transcripts, generation of RNA processing substrates, generation of antisense RNA |
Quantity | 1 System |
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
PR-P1430
|
Promega
P1430 |
500U RNA Polymerase (T3), 5μg pGEM™ express positive control template, 500μL transcription optimized 5X Buffer, 2500U recombinant RNasin™ RNase inhibitor, 100mM 100 |
Each for $324.13
|
|
|||||
PR-P1420
|
Promega
P1420 |
500U RNA Polymerase (SP6), 5μg pGEM™ express positive control template, 500μL transcription optimized 5X Buffer, 2500U recombinant RNasin™ RNase inhibitor, 100mM 100 |
Each for $324.13
|
|
|||||
PR-P1440
|
Promega
P1440 |
500U RNA Polymerase (T7), 5μg pGEM™ express positive control template, 500μL transcription optimized 5X Buffer, 2500U recombinant RNasin™ RNase inhibitor, 100mM 100 |
Each for $324.13
|
|
|||||
Description
- Produce High-Specific-Activity RNA Probes or Defined RNA Transcripts
- Systems available with SP6 RNA Polymerase, T7 RNA Polymerase or T3 RNA Polymerase
- SP6, T7 and T3 RNA Polymerases are extremely promoter-specific, allowing production of virtually homogeneous RNA using plasmid DNA as a template
- Allow synthesis of RNA corresponding to the coding or noncoding strand of cloned DNA from a single plasmid construct
- U.S. Patent 5,552,302; Australian Patent 646803 and other patents.
- Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
Specifications
Store at -20°C. | |
Generation of high-specific-activity RNA probes, generation of microgram quantities of RNA transcripts, generation of RNA processing substrates, generation of antisense RNA | |
1 System |