Epredia™ HistoGel™ Specimen Processing Gel
Encapsulate and retain even small, friable and viscous specimens during histological processing with HistoGel™ Specimen Processing Gel.
Supplier:  Epredia™ HG4000144
 View more versions of this product
Catalog No. 22-045381
Explore available promotions
Description
Description
  • Penetrates specimen without allowing specimen to escape from gel
  • Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
  • Eliminates unwanted discoloration around specimens on slides
  • Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
Specifications
Specifications
Vial
Specimen Processing Gel
144 Pk.