Epredia™ HistoGel™ Specimen Processing Gel
Encapsulate and retain even small, friable and viscous specimens during histological processing with HistoGel™ Specimen Processing Gel.
$454.84 - $5045.91
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | ||||
22-110-678
|
Epredia™
HG4000012 |
Specimen Processing Gel | 12 Pk. |
Case of 12 Each for $454.84
|
|
||||
22-045381
|
Epredia™
HG4000144 |
Specimen Processing Gel | 144 Pk. |
Case of 144 Each for $5,045.91
|
|
||||
Description
- Penetrates specimen without allowing specimen to escape from gel
- Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
- Eliminates unwanted discoloration around specimens on slides
- Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
Specifications
Specimen Processing Gel |