Epredia™ Xylene
Obtain superior lipid extraction, optimal paraffin infiltration, and excellent deparaffinization and clearing during staining with this Xylene.
Supplier:  Epredia™ SH6655
 View more versions of this product
Catalog No. 22-110-618
Explore available promotions
Description
Description
  • For use in all tissue processors and automatic stainers
  • Contains the lowest level of benzene, a known carcinogen
  • Strict quality assurance, including gas chromatography, is performed on each lot to guarantee a consistent product
Specifications
Specifications
55 gallon; steel drum