Epredia™ Xylene
Obtain superior lipid extraction, optimal paraffin infiltration, and excellent deparaffinization and clearing during staining with this Xylene.
$144.65 - $2558.32
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
36-102-5356
|
Epredia™
6601 |
1 Gallon; 4/Cs. |
Case of 4 Each for $408.00
|
|
|||||
66-15
|
Epredia™
6615 |
5 liter; 4/Cs. |
Case of 4 Each for $514.28
|
|
|||||
22-110-618
|
Epredia™
SH6655 |
55 gallon; steel drum |
Each for $2,558.32
|
|
|||||
99-905-01
|
Epredia™
9990501 |
1 Gallon, Ea. |
Each for $144.65
|
|
|||||
Description
- For use in all tissue processors and automatic stainers
- Contains the lowest level of benzene, a known carcinogen
- Strict quality assurance, including gas chromatography, is performed on each lot to guarantee a consistent product