Epredia™ Xylene
Obtain superior lipid extraction, optimal paraffin infiltration, and excellent deparaffinization and clearing during staining with this Xylene.
Supplier:  Epredia™ 6601
 View more versions of this product
Catalog No. 36-102-5356
Explore available promotions
Description
Description
  • For use in all tissue processors and automatic stainers
  • Contains the lowest level of benzene, a known carcinogen
  • Strict quality assurance, including gas chromatography, is performed on each lot to guarantee a consistent product
Specifications
Specifications
1 Gallon; 4/Cs.