Rocks and Minerals Slide Sets
Identify forms, colors, refraction and fossil inclusions.
$851.79 - $851.79
Specifications
Type | Rock and Rock-Forming Minerals |
---|
Includes: Set 1: Basalt, gabbro, gneiss, granite, limestone fossilized, marble, micaschist, quartzite, sandstone, and syenite. Set 2: Andesite, chalk, coal, diorite, limestone oolithic, microgranite, millstone, schist, thyolite, and trachyte.
Description
- Selected rocks and minerals are ground and polished to a thickness of 20 to 30μm
- Mounted with Canada balsam on 1.77 × 1.89 in. (45 × 48mm) slides
- Observe with any normal microscope
- Viewing with polarized light reveals additional detail
Specifications
Rock and Rock-Forming Minerals |