Rocks and Minerals Slide Sets
Identify forms, colors, refraction and fossil inclusions.
Supplier:  Customer Channels Group (Ccg) 7920
 View more versions of this product
Catalog No. S64948
Explore available promotions
Includes: Set 1: Basalt, gabbro, gneiss, granite, limestone fossilized, marble, micaschist, quartzite, sandstone, and syenite. Set 2: Andesite, chalk, coal, diorite, limestone oolithic, microgranite, millstone, schist, thyolite, and trachyte.
Description
Description
  • Selected rocks and minerals are ground and polished to a thickness of 20 to 30μm
  • Mounted with Canada balsam on 1.77 × 1.89 in. (45 × 48mm) slides
  • Observe with any normal microscope
  • Viewing with polarized light reveals additional detail
Specifications
Specifications
Set 1
Rock and Rock-Forming Minerals