missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rosin, gum, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 419175000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentSpecifications
Rosin, gum | |
8050-09-7 | |
Yellow | |
Authentic | |
C20H30O2 | |
500 g | |
4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid | |
RSWGJHLUYNHPMX-UHFFFAOYNA-N | |
Crystalline Chunks and Powder |
163 to 173mg KOH/g | |
75.0°C to 81.0°C | |
180°C | |
Glass bottle | |
MFCD00132205 | |
14,8266 | |
Solubility in water: 130mg/l (20°C) | |
302.46 |
Chemical Identifiers
8050-09-7 | |
302.46 | |
RSWGJHLUYNHPMX-UHFFFAOYNA-N |
C20H30O2 | |
MFCD00132205 | |
4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 232-475-7
RTECSNumber : VL0480000
TSCA : TSCA
RUO – Research Use Only