missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rosin, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03673430

 View more versions of this product

Catalog No. AA3673430


Explore available promotions

Description

Description

Rosin is an ingredient in printing inks, photocopying and laser printing paper, varnishes, adhesives (glues), soap, paper sizing, soda, soldering fluxes and sealing wax. It can be used as a glazing agent in medicines and chewing gum. In industry, rosin is a flux used in soldering. A mixture of pitch and rosin is used to make a surface against which glass is polished when making optical components such as lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Rosin
C20H30O2
250 g
4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid
RSWGJHLUYNHPMX-UHFFFAOYNA-N
Powder or Solid
8050-09-7
MFCD00132205
14,8266
Soluble in alcohol, benzene, ether; Insoluble in water
302.46

Chemical Identifiers

8050-09-7
302.46
RSWGJHLUYNHPMX-UHFFFAOYNA-N
C20H30O2
MFCD00132205
4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

EINECSNumber : 232-475-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only