missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rosin, Thermo Scientific Chemicals

$69.03 - $166.92

Chemical Identifiers

CAS 8050-09-7
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD00132205
InChI Key RSWGJHLUYNHPMX-UHFFFAOYNA-N
Synonym 4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AA3673430
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
03673430
250 g
Each for $69.03
 
AA36734A1
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
036734A1
1 kg
Each for $166.92
 
Description

Description

Rosin is an ingredient in printing inks, photocopying and laser printing paper, varnishes, adhesives (glues), soap, paper sizing, soda, soldering fluxes and sealing wax. It can be used as a glazing agent in medicines and chewing gum. In industry, rosin is a flux used in soldering. A mixture of pitch and rosin is used to make a surface against which glass is polished when making optical components such as lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8050-09-7
302.46
RSWGJHLUYNHPMX-UHFFFAOYNA-N
C20H30O2
MFCD00132205
4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid

Specifications

8050-09-7
MFCD00132205
14,8266
Soluble in alcohol, benzene, ether; Insoluble in water
302.46
Rosin
C20H30O2
250 g
4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid
RSWGJHLUYNHPMX-UHFFFAOYNA-N
Powder or Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

EINECSNumber : 232-475-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only