BUCHI Rotavapor R-100 Rotary Evaporator
The Rotavapor R-100 meets all of your essential needs in evaporation. Features include a manual lift, 4L bath capacity, and digital temperature display.
Supplier:  BUCHI 11100C212
 View more versions of this product
Catalog No. 05-405-106
Explore available promotions
Description
Description
  • Effective Rotavapor for limited budgets
  • Fundamental design fulfills essential needs of evaporation
  • High longevity due to high quality material
  • Reliable operation due to robust design
  • Quick and ergonomic operation with smooth manual lift mechanism
  • Easy flask handling and vapor duct removal due to multifunctional Combi-Clip
  • Easy operation by integrating interface, vacuum pump and recirculating chiller
  • Expandable with wide range of accessories and various glass assemblies
  • Flexibility to use independent heating bath for further applications
Specifications
Specifications
898 mm
100/120 V
4 L
20°C to 95°C
plastic coating P+G
100/120 V 50/60 Hz
Metal, Glass
50/60 Hz
502 mm
617 mm
20 to 280 rpm
Distillation, Concentration, Drying in Pharmaceutical and Reflux Reaction, Re-crystallization, Sublimation in QC and R and D laboratories
Rotavapor, bath and glassware
Cold trap
Rotary Evaporator
R-100