missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rotenone 95.0+%, TCI America™

$237.00 - $788.03

Chemical Identifiers

CAS 83-79-4
Molecular Formula C23H22O6
Molecular Weight (g/mol) 394.423
MDL Number MFCD09025614
InChI Key JUVIOZPCNVVQFO-HBGVWJBISA-N
Synonym rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root
PubChem CID 6758
ChEBI CHEBI:28201
SMILES CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R00905G
View Documents
TCI America
R00905G
5 g
Each for $237.00
 
R009025G
View Documents
TCI America
R009025G
25 g
Each for $788.03
 
Description

Description

Specifications

Chemical Identifiers

83-79-4
394.423
JUVIOZPCNVVQFO-HBGVWJBISA-N
6758
CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC
C23H22O6
MFCD09025614
rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root
CHEBI:28201

Specifications

83-79-4
220°C
MFCD09025614
2811
JUVIOZPCNVVQFO-HBGVWJBISA-N
394.423
CHEBI:28201
≥95.0% (HPLC)
Rotenone
183°C
C23H22O6
5 g
rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root
CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC
6758
394.42
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DJ2800000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'