missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Rotenone | |
183°C | |
C23H22O6 | |
5 g | |
rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root | |
CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC | |
6758 | |
394.42 | |
Crystalline Powder |
83-79-4 | |
220°C | |
MFCD09025614 | |
2811 | |
JUVIOZPCNVVQFO-HBGVWJBISA-N | |
394.423 | |
CHEBI:28201 | |
≥95.0% (HPLC) |
Chemical Identifiers
83-79-4 | |
394.423 | |
JUVIOZPCNVVQFO-HBGVWJBISA-N | |
6758 | |
CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC |
C23H22O6 | |
MFCD09025614 | |
rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root | |
CHEBI:28201 |
Safety and Handling
RTECSNumber : DJ2800000
TSCA : No