Eppendorf™ Rotor for Centrifuge 5910 Ri
Eppendorf Rotor for Centrifuge 5910 Ri-precision in refrigerated centrifugation. Tailor your experiments with varied capacities and speeds, adaptable buckets and adaptors for efficient sample processing. Ideal for temperature-sensitive applications, ensuring accuracy and reproducibility.
Supplier:  Eppendorf™ 5895171001
 View more versions of this product
Catalog No. 13864423
Explore available promotions
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
Specifications
Specifications
S-4x500 - without buckets
4 x 500 mL
4,000 rpm
Without buckets
Aerosol-tight caps available
Rotor
3220 x g
Centrifuge 5910 Ri
4