missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium(III) chloride hydrate, 35 - 40% Ru, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 195480010

 View more versions of this product

Catalog No. AC195480010


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ruthenium(III) chloride hydrate
Black to Brown
Powder
Cl3Ru
MFCD00149844
14, 8302
Solubility in water: soluble
[Cl-].[Cl-].[Cl-].[Ru+3]
207.42
35 to 40% Ru
35 - 40% Ru
14898-67-0
1 g
RuCl3·xH2O
04,421; 08,437; 10,343; 13,268; 14,271; 15,280; 16,292
Ruthenium trichloride
YBCAZPLXEGKKFM-UHFFFAOYSA-K
ruthenium(3+) trichloride
207.42
Glass bottle

Chemical Identifiers

14898-67-0
207.42
YBCAZPLXEGKKFM-UHFFFAOYSA-K
ruthenium(3+) trichloride
Cl3Ru
MFCD00149844
Ruthenium trichloride
[Cl-].[Cl-].[Cl-].[Ru+3]
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May be corrosive to metals.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only