Learn More
Ruthenium(III) chloride hydrate, 35 - 40% Ru, Thermo Scientific Chemicals
$91.90 - $2,676.87
Chemical Identifiers
CAS | 14898-67-0 |
---|---|
Molecular Formula | Cl3Ru |
Molecular Weight (g/mol) | 207.42 |
MDL Number | MFCD00149844 |
InChI Key | YBCAZPLXEGKKFM-UHFFFAOYSA-K |
Synonym | Ruthenium trichloride |
IUPAC Name | ruthenium(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Ru+3] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC195480010
|
Thermo Scientific Chemicals
195480010 |
1 g | Glass bottle |
N/A
|
|
||||
AC195480050
|
Thermo Scientific Chemicals
195480050 |
5 g | Glass bottle |
N/A
|
|
||||
AC195480250
|
Thermo Scientific Chemicals
195480250 |
25 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14898-67-0 | |
207.42 | |
YBCAZPLXEGKKFM-UHFFFAOYSA-K | |
ruthenium(3+) trichloride |
Cl3Ru | |
MFCD00149844 | |
Ruthenium trichloride | |
[Cl-].[Cl-].[Cl-].[Ru+3] |
Specifications
Black to Brown | |
Powder | |
Cl3Ru | |
MFCD00149844 | |
14, 8302 | |
Solubility in water: soluble | |
[Cl-].[Cl-].[Cl-].[Ru+3] | |
207.42 | |
35 to 40% Ru | |
Ruthenium(III) chloride hydrate, 35 - 40% Ru |
14898-67-0 | |
1 g | |
RuCl3·xH2O | |
04,421; 08,437; 10,343; 13,268; 14,271; 15,280; 16,292 | |
Ruthenium trichloride | |
YBCAZPLXEGKKFM-UHFFFAOYSA-K | |
ruthenium(3+) trichloride | |
207.42 | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May be corrosive to metals.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO – Research Use Only