Learn More
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate, 95%, Thermo Scientific Chemicals
$83.63 - $231.92
Chemical Identifiers
CAS | 73671-79-1 |
---|---|
Molecular Formula | C13H11NO |
Molecular Weight (g/mol) | 197.24 |
MDL Number | MFCD00064181 |
InChI Key | GONOHGQPZFXJOJ-UHFFFAOYNA-N |
Synonym | s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate |
PubChem CID | 7023510 |
IUPAC Name | 1-[(1S)-1-isocyanatoethyl]naphthalene |
SMILES | CC(N=C=O)C1=C2C=CC=CC2=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
AAL20242MD
|
Thermo Scientific Chemicals
L20242MD |
250 mg |
Each for $83.63
|
|
|||||
AAL2024203
|
Thermo Scientific Chemicals
L2024203 |
1 g |
Each for $231.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
73671-79-1 | |
197.24 | |
GONOHGQPZFXJOJ-UHFFFAOYNA-N | |
7023510 | |
CC(N=C=O)C1=C2C=CC=CC2=CC=C1 |
C13H11NO | |
MFCD00064181 | |
s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate | |
1-[(1S)-1-isocyanatoethyl]naphthalene |
Specifications
73671-79-1 | |
106°C to 108°C (0.16 mmHg) | |
C13H11NO | |
MFCD00064181 | |
UN2206 | |
Moisture sensitive; May form a precipitate | |
GONOHGQPZFXJOJ-UHFFFAOYNA-N | |
CC(N=C=O)C1=C2C=CC=CC2=CC=C1 | |
197.24 | |
197.24 | |
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate |
1.118 | |
93°C (199°F) | |
1.6045 | |
250 mg | |
4675069 | |
s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate | |
+47° (c=3.5 in Toluene) | |
1-[(1S)-1-isocyanatoethyl]naphthalene | |
7023510 | |
95% |
Safety and Handling
GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only