Learn More
(S)-(+)-1-Bromo-2-methylbutane, 97%, stab. with potassium carbonate, Thermo Scientific Chemicals
$269.26 - $886.30
Chemical Identifiers
CAS | 534-00-9 |
---|---|
Molecular Formula | C5H11Br |
Molecular Weight (g/mol) | 151.05 |
MDL Number | MFCD00078662 |
InChI Key | XKVLZBNEPALHIO-YFKPBYRVSA-N |
Synonym | s-1-bromo-2-methylbutane, s-+-1-bromo-2-methylbutane, butane, 1-bromo-2-methyl-, 2s, unii-cgs982pbtw, 2s-1-bromo-2-methylbutane, d-amyl bromide, cgs982pbtw, 2s-1-bromo-2-methyl-butane, +-2-methylbutyl bromide, butane, 1-bromo-2-methyl-, s |
PubChem CID | 5464167 |
IUPAC Name | (2S)-1-bromo-2-methylbutane |
SMILES | CC[C@H](C)CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2707806
|
Thermo Scientific Chemicals
H2707806 |
5 g |
Each for $269.26
|
|
|||||
AAH2707814
|
Thermo Scientific Chemicals
H2707814 |
25 g |
Each for $886.30
|
|
|||||
Description
(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases. Light sensitive.
Chemical Identifiers
534-00-9 | |
151.05 | |
XKVLZBNEPALHIO-YFKPBYRVSA-N | |
5464167 | |
CC[C@H](C)CBr |
C5H11Br | |
MFCD00078662 | |
s-1-bromo-2-methylbutane, s-+-1-bromo-2-methylbutane, butane, 1-bromo-2-methyl-, 2s, unii-cgs982pbtw, 2s-1-bromo-2-methylbutane, d-amyl bromide, cgs982pbtw, 2s-1-bromo-2-methyl-butane, +-2-methylbutyl bromide, butane, 1-bromo-2-methyl-, s | |
(2S)-1-bromo-2-methylbutane |
Specifications
534-00-9 | |
121°C to 122°C | |
C5H11Br | |
MFCD00078662 | |
UN1993 | |
Light sensitive; May form a precipitate | |
s-1-bromo-2-methylbutane, s-+-1-bromo-2-methylbutane, butane, 1-bromo-2-methyl-, 2s, unii-cgs982pbtw, 2s-1-bromo-2-methylbutane, d-amyl bromide, cgs982pbtw, 2s-1-bromo-2-methyl-butane, +-2-methylbutyl bromide, butane, 1-bromo-2-methyl-, s | |
XKVLZBNEPALHIO-YFKPBYRVSA-N | |
CC[C@H](C)CBr | |
151.05 | |
151.05 | |
(S)-(+)-1-Bromo-2-methylbutane, stabilized with potassium carbonate |
1.223 | |
22°C (79°F) | |
1.4455 | |
5 g | |
1718815 | |
14,601 | |
Insoluble in water. | |
+4.5° (Neat) | |
(2S)-1-bromo-2-methylbutane | |
5464167 | |
97% |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 208-583-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only