Learn More
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0985514
Description
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid | |
154°C to 156°C | |
C14H14O3 | |
25 g | |
3591067 | |
naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno | |
CMWTZPSULFXXJA-VIFPVBQESA-N | |
CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O | |
230.263 | |
CHEBI:7476 | |
99% |
22204-53-1 | |
Odorless | |
MFCD00010500 | |
UN2811 | |
14,6417 | |
Insoluble in water. | |
+66° (c=1 in Chloroform) | |
(2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid | |
156391 | |
230.26 |
Chemical Identifiers
22204-53-1 | |
230.263 | |
CMWTZPSULFXXJA-VIFPVBQESA-N | |
156391 | |
(2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid |
C14H14O3 | |
MFCD00010500 | |
naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno | |
CHEBI:7476 | |
CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 244-838-7
RTECSNumber : UF5275000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only