Learn More
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%, Thermo Scientific Chemicals
$53.44 - $363.00
Chemical Identifiers
CAS | 22204-53-1 |
---|---|
Molecular Formula | C14H14O3 |
Molecular Weight (g/mol) | 230.263 |
MDL Number | MFCD00010500 |
InChI Key | CMWTZPSULFXXJA-VIFPVBQESA-N |
Synonym | naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno |
PubChem CID | 156391 |
ChEBI | CHEBI:7476 |
IUPAC Name | (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid |
SMILES | CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0985503
|
Thermo Scientific Chemicals
L0985503 |
1 g |
Each for $53.44
|
|
|||||
AAL0985506
|
Thermo Scientific Chemicals
L0985506 |
5 g |
Each for $145.79
|
|
|||||
AAL0985514
|
Thermo Scientific Chemicals
L0985514 |
25 g |
Each for $363.00
|
|
|||||
Description
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
22204-53-1 | |
230.263 | |
CMWTZPSULFXXJA-VIFPVBQESA-N | |
156391 | |
(2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid |
C14H14O3 | |
MFCD00010500 | |
naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno | |
CHEBI:7476 | |
CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O |
Specifications
22204-53-1 | |
Odorless | |
MFCD00010500 | |
UN2811 | |
14,6417 | |
Insoluble in water. | |
+66° (c=1 in Chloroform) | |
(2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid | |
156391 | |
230.26 | |
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid |
154°C to 156°C | |
C14H14O3 | |
1 g | |
3591067 | |
naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno | |
CMWTZPSULFXXJA-VIFPVBQESA-N | |
CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O | |
230.263 | |
CHEBI:7476 | |
99% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 244-838-7
RTECSNumber : UF5275000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only