Learn More
(S)-(-)-3-Butyn-2-ol, 95%, 98% ee, Thermo Scientific™
Chemical Identifiers
CAS | 2914-69-4 |
---|---|
Molecular Formula | C4H6O |
Molecular Weight (g/mol) | 70.09 |
MDL Number | MFCD00190166 |
InChI Key | GKPOMITUDGXOSB-BYPYZUCNSA-N |
Synonym | s---3-butyn-2-ol, s-3-butyn-2-ol, 2s-but-3-yn-2-ol, s-but-3-yn-2-ol, 3-butyn-2-ol, 2s, s---1-butyn-3-ol, 2s-3-butyn-2-ol, 3r-+-3-hydroxybut-1-yne, s---3-butyn-2-ol, purum gc |
PubChem CID | 6995470 |
IUPAC Name | (2S)-but-3-yn-2-ol |
SMILES | CC(C#C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC362410010
|
Thermo Scientific Chemicals
362410010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC362410050
|
Thermo Scientific Chemicals
362410050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2914-69-4 | |
70.09 | |
GKPOMITUDGXOSB-BYPYZUCNSA-N | |
6995470 | |
CC(C#C)O |
C4H6O | |
MFCD00190166 | |
s---3-butyn-2-ol, s-3-butyn-2-ol, 2s-but-3-yn-2-ol, s-but-3-yn-2-ol, 3-butyn-2-ol, 2s, s---1-butyn-3-ol, 2s-3-butyn-2-ol, 3r-+-3-hydroxybut-1-yne, s---3-butyn-2-ol, purum gc | |
(2S)-but-3-yn-2-ol |
Specifications
2914-69-4 | |
100.0 | |
108°C to 111°C | |
97.5% min. (GC) | |
94% min. (GC) | |
C4H6O | |
1g | |
s---3-butyn-2-ol, s-3-butyn-2-ol, 2s-but-3-yn-2-ol, s-but-3-yn-2-ol, 3-butyn-2-ol, 2s, s---1-butyn-3-ol, 2s-3-butyn-2-ol, 3r-+-3-hydroxybut-1-yne, s---3-butyn-2-ol, purum gc | |
CC(C#C)O | |
70.09 | |
70.09 | |
(S)-(-)-3-Butyn-2-ol, 95%, (98% e.e.) |
94.0 | |
0.8940g/mL | |
26°C | |
Authentic | |
Glass bottle | |
MFCD00190166 | |
0.894 | |
GKPOMITUDGXOSB-BYPYZUCNSA-N | |
(2S)-but-3-yn-2-ol | |
6995470 | |
95%,98% ee |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Fatal in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Fatal if swallowed.
Flammable liquid and vapor.
Fatal if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
TSCA : TSCA
RUO â Research Use Only