missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2243514

 View more versions of this product

Catalog No. AAB2243514


Explore available promotions

Description

Description

S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications

Specifications

S-Boc-2-mercapto-4,6-dimethylpyrimidine
48°C to 51°C
C11H16N2O2S
25 g
s-boc-2-mercapto-4,6-dimethylpyrimidine, ccris 2600, 2-tert-butoxycarbonylthio-4,6-dimethylpyrimidine, 2-boc-thio-4,6-dimethylpyrimidine, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiocarbonate, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiolcarbonate, o-tert-butyl s-4,6-dimethylpyrimidin-2-yl carbonothioate, carbonothioic acid, o-1,1-dimethylethyl s-4,6-dimethyl-2-pyrimidinyl ester, tert-butyl 4,6-dimethylpyrimidin-2-yl sulfanylformate, thiocarbonic acid, o-tert-butyl s-4,6-dimethyl-2-pyrimidinyl ester
CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1
240.32
240.33
41840-28-2
>110°C (230°F)
MFCD00006080
396984
POTDIELOEHTPJN-UHFFFAOYSA-N
tert-butyl (4,6-dimethylpyrimidin-2-yl)sulfanylformate
148428
97%

Chemical Identifiers

41840-28-2
240.32
POTDIELOEHTPJN-UHFFFAOYSA-N
148428
CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1
C11H16N2O2S
MFCD00006080
s-boc-2-mercapto-4,6-dimethylpyrimidine, ccris 2600, 2-tert-butoxycarbonylthio-4,6-dimethylpyrimidine, 2-boc-thio-4,6-dimethylpyrimidine, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiocarbonate, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiolcarbonate, o-tert-butyl s-4,6-dimethylpyrimidin-2-yl carbonothioate, carbonothioic acid, o-1,1-dimethylethyl s-4,6-dimethyl-2-pyrimidinyl ester, tert-butyl 4,6-dimethylpyrimidin-2-yl sulfanylformate, thiocarbonic acid, o-tert-butyl s-4,6-dimethyl-2-pyrimidinyl ester
tert-butyl (4,6-dimethylpyrimidin-2-yl)sulfanylformate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 255-562-1

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only