missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%, Thermo Scientific Chemicals
$75.93 - $254.18
Chemical Identifiers
CAS | 41840-28-2 |
---|---|
Molecular Formula | C11H16N2O2S |
Molecular Weight (g/mol) | 240.32 |
MDL Number | MFCD00006080 |
InChI Key | POTDIELOEHTPJN-UHFFFAOYSA-N |
Synonym | s-boc-2-mercapto-4,6-dimethylpyrimidine, ccris 2600, 2-tert-butoxycarbonylthio-4,6-dimethylpyrimidine, 2-boc-thio-4,6-dimethylpyrimidine, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiocarbonate, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiolcarbonate, o-tert-butyl s-4,6-dimethylpyrimidin-2-yl carbonothioate, carbonothioic acid, o-1,1-dimethylethyl s-4,6-dimethyl-2-pyrimidinyl ester, tert-butyl 4,6-dimethylpyrimidin-2-yl sulfanylformate, thiocarbonic acid, o-tert-butyl s-4,6-dimethyl-2-pyrimidinyl ester |
PubChem CID | 148428 |
IUPAC Name | tert-butyl (4,6-dimethylpyrimidin-2-yl)sulfanylformate |
SMILES | CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2243506
|
Thermo Scientific Chemicals
B2243506 |
5 g |
Each for $75.93
|
|
|||||
AAB2243514
|
Thermo Scientific Chemicals
B2243514 |
25 g |
Each for $254.18
|
|
|||||
Description
S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsS-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Chemical Identifiers
41840-28-2 | |
240.32 | |
POTDIELOEHTPJN-UHFFFAOYSA-N | |
148428 | |
CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1 |
C11H16N2O2S | |
MFCD00006080 | |
s-boc-2-mercapto-4,6-dimethylpyrimidine, ccris 2600, 2-tert-butoxycarbonylthio-4,6-dimethylpyrimidine, 2-boc-thio-4,6-dimethylpyrimidine, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiocarbonate, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiolcarbonate, o-tert-butyl s-4,6-dimethylpyrimidin-2-yl carbonothioate, carbonothioic acid, o-1,1-dimethylethyl s-4,6-dimethyl-2-pyrimidinyl ester, tert-butyl 4,6-dimethylpyrimidin-2-yl sulfanylformate, thiocarbonic acid, o-tert-butyl s-4,6-dimethyl-2-pyrimidinyl ester | |
tert-butyl (4,6-dimethylpyrimidin-2-yl)sulfanylformate |
Specifications
41840-28-2 | |
>110°C (230°F) | |
MFCD00006080 | |
396984 | |
POTDIELOEHTPJN-UHFFFAOYSA-N | |
tert-butyl (4,6-dimethylpyrimidin-2-yl)sulfanylformate | |
148428 | |
97% |
48°C to 51°C | |
C11H16N2O2S | |
5 g | |
s-boc-2-mercapto-4,6-dimethylpyrimidine, ccris 2600, 2-tert-butoxycarbonylthio-4,6-dimethylpyrimidine, 2-boc-thio-4,6-dimethylpyrimidine, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiocarbonate, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiolcarbonate, o-tert-butyl s-4,6-dimethylpyrimidin-2-yl carbonothioate, carbonothioic acid, o-1,1-dimethylethyl s-4,6-dimethyl-2-pyrimidinyl ester, tert-butyl 4,6-dimethylpyrimidin-2-yl sulfanylformate, thiocarbonic acid, o-tert-butyl s-4,6-dimethyl-2-pyrimidinyl ester | |
CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1 | |
240.32 | |
240.33 | |
S-Boc-2-mercapto-4,6-dimethylpyrimidine |
Safety and Handling
EINECSNumber : 255-562-1
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only