missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-Carboxymethyl-L-cysteine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2348714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
∼200°C (decomposition) | |
638-23-3 | |
MFCD00002614 | |
14,1802 | |
GBFLZEXEOZUWRN-VKHMYHEASA-N | |
(2R)-2-amino-3-(carboxymethylsulfanyl)propanoic acid | |
179.19 | |
CHEBI:16163 | |
179.2 |
S-Carboxymethyl-L-cysteine | |
C5H9NO4S | |
1725012 | |
s-carboxymethyl-l-cysteine, carbocisteine, carbocysteine, carbocistein, transbronchin, bronchokod, lisomucil, mucolase, mucotab | |
N[C@@H](CSCC(O)=O)C(O)=O | |
25 g | |
193653 | |
−29° (c=10 in 10% NaHCO) | |
97% |
Chemical Identifiers
638-23-3 | |
179.19 | |
GBFLZEXEOZUWRN-VKHMYHEASA-N | |
193653 | |
N[C@@H](CSCC(O)=O)C(O)=O |
C5H9NO4S | |
MFCD00002614 | |
s-carboxymethyl-l-cysteine, carbocisteine, carbocysteine, carbocistein, transbronchin, bronchokod, lisomucil, mucolase, mucotab | |
CHEBI:16163 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-327-5
RTECSNumber : AY4342000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only