missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-Glycidyl nosylate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 296420025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(S)-(+)-Glycidyl nosylate | |
Authentic | |
Glass bottle | |
MFCD00064582 | |
+ 21.50 (20.00°C c=2,CHCl3) | |
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate | |
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1 | |
259.23 | |
259.23 |
115314-14-2 | |
97.5% min. (GC) | |
C9H9NO6S | |
2.5g | |
+ 21.50 | |
AIHIHVZYAAMDPM-QMMMGPOBSA-N | |
[(2S)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate | |
146490 | |
98% |
Chemical Identifiers
115314-14-2 | |
259.23 | |
AIHIHVZYAAMDPM-QMMMGPOBSA-N | |
146490 | |
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1 |
C9H9NO6S | |
MFCD00064582 | |
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate | |
[(2S)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only