missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-Glycidyl nosylate, 98%, Thermo Scientific™

Chemical Identifiers

CAS 115314-14-2
Molecular Formula C9H9NO6S
Molecular Weight (g/mol) 259.23
MDL Number MFCD00064582
InChI Key AIHIHVZYAAMDPM-QMMMGPOBSA-N
Synonym s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
PubChem CID 146490
IUPAC Name [(2S)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate
SMILES [O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC296420025
SDS View Documents
Thermo Scientific Chemicals
296420025
2.5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC296420100
SDS View Documents
Thermo Scientific Chemicals
296420100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC296420500
SDS View Documents
Thermo Scientific Chemicals
296420500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

115314-14-2
259.23
AIHIHVZYAAMDPM-QMMMGPOBSA-N
146490
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1
C9H9NO6S
MFCD00064582
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
[(2S)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate

Specifications

115314-14-2
97.5% min. (GC)
C9H9NO6S
2.5g
+ 21.50
AIHIHVZYAAMDPM-QMMMGPOBSA-N
[(2S)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate
146490
98%
Authentic
Glass bottle
MFCD00064582
+ 21.50 (20.00°C c=2,CHCl3)
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1
259.23
259.23
(S)-(+)-Glycidyl nosylate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only