missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-Mandelic acid, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1506114

 View more versions of this product

Catalog No. AAA1506114


Explore available promotions

Description

Description

(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.

Solubility
Soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

(S)-(+)-Mandelic acid
17199-29-0
MFCD00004495
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
IWYDHOAUDWTVEP-ZETCQYMHSA-N
(2S)-2-hydroxy-2-phenylacetic acid
439616
152.15
25 g
Light sensitive
131°C to 135°C
C8H8O3
2208678
Soluble in water.
C1=CC=C(C=C1)C(C(=O)O)O
152.149
CHEBI:32800
≥99%
+155° (c=5 in Water)

Chemical Identifiers

17199-29-0
152.149
IWYDHOAUDWTVEP-ZETCQYMHSA-N
439616
(2S)-2-hydroxy-2-phenylacetic acid
C8H8O3
MFCD00004495
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
CHEBI:32800
C1=CC=C(C=C1)C(C(=O)O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P280-P305+P351+P338-P310

H318

EINECSNumber : 241-240-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only