missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-α-Methoxyphenylacetic acid, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 224490010

 View more versions of this product

Catalog No. AC224490010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chiral derivatization reagent
Specifications

Specifications

(S)-(+)-α-Methoxyphenylacetic acid
98.5
Authentic
Glass bottle
C6H5CH(OCH3)COOH
1g
+160° (20°C c=1,methanol)
s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s
DIWVBIXQCNRCFE-QMMMGPOBSA-N
(2S)-2-methoxy-2-phenylacetic acid
643325
99%
26164-26-1
100.0
99%
C9H10O3
MFCD00064216
10, I, 84
+ 160
Solubility in water: soluble in water
COC(C1=CC=CC=C1)C(=O)O
166.18
166.18

Chemical Identifiers

26164-26-1
166.18
DIWVBIXQCNRCFE-QMMMGPOBSA-N
643325
COC(C1=CC=CC=C1)C(=O)O
C9H10O3
MFCD00064216
s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s
(2S)-2-methoxy-2-phenylacetic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 247-492-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only