missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-α-Methoxyphenylacetic acid, 99%, Thermo Scientific™

Chemical Identifiers

CAS 26164-26-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00064216
InChI Key DIWVBIXQCNRCFE-QMMMGPOBSA-N
Synonym s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s
PubChem CID 643325
IUPAC Name (2S)-2-methoxy-2-phenylacetic acid
SMILES COC(C1=CC=CC=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC224490010
SDS View Documents
Thermo Scientific Chemicals
224490010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC224490050
SDS View Documents
Thermo Scientific Chemicals
224490050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chiral derivatization reagent
Specifications

Chemical Identifiers

26164-26-1
166.18
DIWVBIXQCNRCFE-QMMMGPOBSA-N
643325
COC(C1=CC=CC=C1)C(=O)O
C9H10O3
MFCD00064216
s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s
(2S)-2-methoxy-2-phenylacetic acid

Specifications

26164-26-1
100.0
99%
C9H10O3
MFCD00064216
10, I, 84
+ 160
Solubility in water: soluble in water
COC(C1=CC=CC=C1)C(=O)O
166.18
166.18
(S)-(+)-α-Methoxyphenylacetic acid
98.5
Authentic
Glass bottle
C6H5CH(OCH3)COOH
1g
+160° (20°C c=1,methanol)
s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s
DIWVBIXQCNRCFE-QMMMGPOBSA-N
(2S)-2-methoxy-2-phenylacetic acid
643325
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 247-492-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only