FEATHER Safeshield Scalpel
FEATHER Safeshield scalpel features a clear protective shield that can be retracted or advanced multiple times to cover the blade. Multiple sizes are available with color-coded button on the protective shield to assist in rapid blade size identification.
Supplier: FEATHER SS10
Description
- Made in Japan
- Indispensable in emergencies
- Safety shield can be advanced to cover the blade and retracted multiple times
- Color-coded button on the protective shield for the blade identification
- Individually packed
- Single use only
Specifications
Stainless Steel | |
Single-use | |
ISO13485:2016 |
# 10 | |
Sterile | |
10/Pk. |