FEATHER Safeshield Scalpel
FEATHER Safeshield scalpel features a clear protective shield that can be retracted or advanced multiple times to cover the blade. Multiple sizes are available with color-coded button on the protective shield to assist in rapid blade size identification.
Supplier:  FEATHER SS11
 View more versions of this product
Catalog No. 36-101-5345
Explore available promotions
Description
Description
  • Made in Japan
  • Indispensable in emergencies
  • Safety shield can be advanced to cover the blade and retracted multiple times
  • Color-coded button on the protective shield for the blade identification
  • Individually packed
  • Single use only
Specifications
Specifications
Stainless Steel
Single-use
ISO13485:2016
# 11
Sterile
10/Pk.