Safetec™ Formaldehyde Spill Response Kit DFS Item
Ideal for small spills
Supplier:  Safetec™ 48630
 View more versions of this product
Catalog No. 19-314-634
Explore available promotions
Includes:
Pair of nitrile gloves, respirator, open back gown, vented goggles, FSC-1 solidifier (11oz. bottle), four scoops/scrapers, three wiper pads, four clear disposal bags (30 x 36 in.), four twist ties, instructions, hard vinyl case
Description
Description
Emergency response kit for containing small spills in the lab immobilizes formalin and minimizes off-gassing.
Features FSC-1 to aid in reducing exposure to formaldehyde gas in the event of a spill. The formalin is transformed into an easily removable semi-solid mass and the resulting mass cools rapidly, minimizing formaldehyde off gassing.
Specifications
Specifications
Formaldehyde Spill Response Kit
Small formaldehyde spills
One pair of nitrile gloves, respirator, open back gown, vented goggles, FSC-1 Solidifier (11 oz. bottle), 4 scoop/scrapers, 3 pads, (4) 30 x 36 in. clear disposal bags, 4 twist ties, instructions