Safetec™ Formaldehyde Spill Response Kit
Ideal for small spills
$411.67 - $484.95
Specifications
For Use With (Application) | Small formaldehyde spills |
---|
Includes: Pair of nitrile gloves, respirator, open back gown, vented goggles, FSC-1 solidifier (11oz. bottle), four scoops/scrapers, three wiper pads, four clear disposal bags (30 × 36 in.), four twist ties, instructions, hard vinyl case
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | ||||
19-314-635
|
Safetec™
48625 |
Refill | One pair of nitrile gloves, respirator, open back gown, vented goggles, FSC-1 Solidifier (11 oz. bottle), 4 scoop/scrapers, 3 wiper pads, (4) 30 x 36 in. clear disposal bags, 4 twist ties, instructions |
Each for $411.67
|
|
||||
19-314-634
|
Safetec™
48630 |
Formaldehyde Spill Response Kit | One pair of nitrile gloves, respirator, open back gown, vented goggles, FSC-1 Solidifier (11 oz. bottle), 4 scoop/scrapers, 3 pads, (4) 30 x 36 in. clear disposal bags, 4 twist ties, instructions |
Each for $484.95
|
|
||||
Description
Emergency response kit for containing small spills in the lab immobilizes formalin and minimizes off-gassing.
Features FSC-1 to aid in reducing exposure to formaldehyde gas in the event of a spill. The formalin is transformed into an easily removable semi-solid mass and the resulting mass cools rapidly, minimizing formaldehyde off gassing.
Specifications
Small formaldehyde spills |