Safetec™ Formaldehyde Spill Response Kit DFS Item
Ideal for small spills
Supplier:  Safetec™ 48625
 View more versions of this product
Catalog No. 19-314-635
Explore available promotions
Includes:
Pair of nitrile gloves, respirator, open back gown, vented goggles, FSC-1 solidifier (11oz. bottle), four scoops/scrapers, three wiper pads, four clear disposal bags (30 x 36 in.), four twist ties, instructions, hard vinyl case
Description
Description
Emergency response kit for containing small spills in the lab immobilizes formalin and minimizes off-gassing.
Features FSC-1 to aid in reducing exposure to formaldehyde gas in the event of a spill. The formalin is transformed into an easily removable semi-solid mass and the resulting mass cools rapidly, minimizing formaldehyde off gassing.
Specifications
Specifications
Refill
Small formaldehyde spills
One pair of nitrile gloves, respirator, open back gown, vented goggles, FSC-1 Solidifier (11 oz. bottle), 4 scoop/scrapers, 3 wiper pads, (4) 30 x 36 in. clear disposal bags, 4 twist ties, instructions