Accuform™ Safety Sign - NOTICE TO ENTER EMPLOYEES & VISITORS REQUIRED TO SHOW PROOF OF COVID-19 VACCINATION, Adhesive Vinyl
Accuform™ vaccination signs communicate health and safety policies, reinforce procedures, and identify locations
Supplier:  Accuform™ MBDX808VS
 View more versions of this product
Catalog No. 19-119-102
Explore available promotions
Description
Description
  • Printed matte surface has permanent acrylic adhesive back
  • Flexible vinyl can be applied to curved or uneven surfaces
  • Moisture resistant for various light-duty indoor and sheltered outdoor marking needs
  • Square corners and scored back liner for quick peel-off and application
  • Complies with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Specifications
    Specifications
    Vinyl
    Rectangular
    NOTICE TO ENTER EMPLOYEES & VISITORS REQUIRED TO SHOW PROOF OF COVID-19 VACCINATION
    Self-adhesive
    English
    Blue on White