missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Safranine, 1% (w/v), Counterstain for Gram Staining, Ricca Chemical

Supplier:  Ricca Chemical Company RI66604

 View more versions of this product

Catalog No. 6660-4


Explore available promotions

Description

Description

Specifications

Specifications

Safranine
477-73-6,7732-18-5
100.0,1.01
C20H19ClN4
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
2723800
Aqueous Solution
120 mL
Counterstain for Gram Staining
97.0,0.97
1% (w/v)
MFCD00011759
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
350.85
Natural Poly Bottle
Red
Liquid

Chemical Identifiers

477-73-6
350.85
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
C20H19ClN4
MFCD00011759
2723800
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
Videos
SDS
Documents

Documents

Promotions

Promotions