missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Safranine, 1% (w/v), Counterstain for Gram Staining, Ricca Chemical
$76.31 - $511.78
Chemical Identifiers
CAS | 477-73-6 |
---|---|
Molecular Formula | C20H19ClN4 |
Molecular Weight (g/mol) | 350.85 |
MDL Number | MFCD00011759 |
InChI Key | QRYAEWIQIBAZOJ-UHFFFAOYSA-N |
PubChem CID | 2723800 |
IUPAC Name | 2,7-diamino-1,8-dimethyl-5-phenyl-5λâµ-phenazin-5-ylium chloride |
SMILES | [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6660-4
|
Ricca Chemical Company
RI66604 |
120 mL | Natural Poly Bottle |
Each for $76.31
|
|
||||
6660-16
|
Ricca Chemical Company
666016 |
500 mL | Natural Poly Bottle |
Each for $117.97
|
|
||||
6660-1
|
Ricca Chemical Company
66601 |
4 L | Natural Poly Bottle |
Each for $511.78
|
|
||||
Chemical Identifiers
477-73-6 | |
350.85 | |
QRYAEWIQIBAZOJ-UHFFFAOYSA-N | |
2,7-diamino-1,8-dimethyl-5-phenyl-5λâµ-phenazin-5-ylium chloride |
C20H19ClN4 | |
MFCD00011759 | |
2723800 | |
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1 |
Specifications
477-73-6,7732-18-5 | |
100.0,1.01 | |
C20H19ClN4 | |
QRYAEWIQIBAZOJ-UHFFFAOYSA-N | |
2,7-diamino-1,8-dimethyl-5-phenyl-5λâµ-phenazin-5-ylium chloride | |
2723800 | |
Aqueous Solution | |
120 mL | |
Safranine, Counterstain for Gram Staining |
97.0,0.97 | |
1% (w/v) | |
MFCD00011759 | |
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1 | |
350.85 | |
Natural Poly Bottle | |
Red | |
Liquid |